Anti-FBXW12 Mouse Monoclonal Antibody [clone: 1A9]
Catalog # 103348-170
Supplier: Novus Biologicals
Specifications
- Antibody Type:Primary
- Antigen Symbol:FBXW12
- Clonality:Monoclonal
- Clone:1A9
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:IgG2a kappa
- Reactivity:Human
- Western Blot:Yes
- Size:100 μg
- Gene ID:285231
- Antigen Synonyms:F-box and WD-40 domain-containing protein 12|F-box only protein 35MGC120386|F-box- and WD40-repeat-containing protein|F-box/WD repeat-containing protein 12|FBXO12FBXO35F-box and WD-40 domain protein 12|MGC120385|Fbw12|FBW12MGC120387|F-box and WD repeat domain containing 12
- Storage Buffer:PBS (pH 7.4)
- Storage Temperature:Store at -20C. Avoid freeze-thaw cycles.
- Immunogen:FBXW12 (NP_996985, 265 a.a. ~ 355 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.LLRPSEGSDPLSTFLPHKLCASACWTPKVKNRITLMSQSSTGKKTEFITFDLTTKKTGGQTVIQAYEIASFQVAAHLKCPIWMGASDGYM
- Purification:Protein A purified
- Cat. No.:103348-170
- Supplier no.:H00285231-M03
Specifications
About this item
The FBXW12 Antibody (1A9) from Novus Biologicals is a mouse monoclonal antibody to FBXW12. This antibody reacts with human. The FBXW12 Antibody (1A9) has been validated for the following applications: Western Blot, ELISA.
Type: Primary
Antigen: FBXW12
Clonality: Monoclonal
Clone: 1A9
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a Kappa
Reactivity: Human