Anti-EXOSC3 Mouse Monoclonal Antibody [clone: 3E5]
Catalog # 103343-572
Supplier: Novus Biologicals
Specifications
- Antibody Type:Primary
- Antigen Symbol:EXOSC3
- Clonality:Monoclonal
- Clone:3E5
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- ImmunoPrecipitation:Yes
- Isotype:IgG2a kappa
- Reactivity:Human
- Size:100 μg
- Gene ID:51010
- Antigen Synonyms:hRrp-40|hRrp40p|p10Rrp40p|RP11-3J10.8|Ribosomal RNA-processing protein 40|bA3J10.7|exosome component 3MGC15120|RRP40MGC723|exosome complex exonuclease RRP40|exosome component Rrp40|CGI-102
- Storage Buffer:PBS (pH 7.4)
- Storage Temperature:Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
- Immunogen:EXOSC3 (NP_057126, 176 a.a. ~ 275 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
- Purification:IgG purified
- Cat. No.:103343-572
- Supplier no.:H00051010-M11
Specifications
About this item
The EXOSC3 Antibody (3E5) from Novus Biologicals is a mouse monoclonal antibody to EXOSC3. This antibody reacts with human. The EXOSC3 Antibody (3E5) has been validated for the following applications: ELISA, Immunoprecipitation.
Type: Primary
Antigen: EXOSC3
Clonality: Monoclonal
Clone: 300000
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a Kappa
Reactivity: Human