Anti-EMR3 Armenian Hamster Monoclonal Antibody (PE (Phycoerythrin)) [clone: 3D7]
Catalog # 103406-930
Supplier: Novus Biologicals
Specifications
- Antibody Type:Primary
- Antigen Symbol:EMR3
- Clonality:Monoclonal
- Clone:3D7
- Conjugation:PE (Phycoerythrin)
- Flow Cytometry:Yes
- Host:Armenian hamster
- Isotype:IgG
- Reactivity:Human
- Size:100 μl
- Gene ID:84658
- Antigen Synonyms:egf-like module containing|EGF-like module-containing mucin-like hormone receptor-like 3|egf-like module-containing mucin-like receptor 3|EGF-like module receptor 3|mucin-like|hormone receptor-like 3
- Storage Buffer:PBS
- Storage Temperature:Store at 4C in the dark.
- Immunogen:ARHO-EMR3-CD97 (EGF1) transfectants.
- Purification:Protein G purified
- Cat. No.:103406-930
Specifications
About this item
The EMR3 Antibody (3D7) [PE] from Novus Biologicals is a armenian hamster monoclonal antibody to EMR3. This antibody reacts with human. The EMR3 Antibody (3D7) [PE] has been validated for the following applications: Flow Cytometry.
Type: Primary
Antigen: EMR3
Clonality: Monoclonal
Clone: 3D7
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Armenian Hamster
Isotype: IgG
Reactivity: Human