To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Antibody Type:Primary
- Antigen Symbol:EGLN1/PHD2
- Clonality:Polyclonal
- Conjugation:DyLight® 405
- Flow Cytometry:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Primates,Human,Rat,Mouse
- Western Blot:Yes
- Size:100 μl
- Environmentally Preferable:
- Gene ID:54583
- Antigen Synonyms:HIFPH2 EGL nine (C.elegans) homolog 1|ZMYND6DKFZp761F179|EC 1.14.11.-|HPH-2|Hypoxia-inducible factor prolyl hydroxylase 2|egl nine homolog 1|egl nine homolog 1 (C. elegans)|Prolyl hydroxylase domain-containing protein 2|egl nine-like protein 1|HIF prolyl hydroxylase 2|HIF-PH2|C1orf12HPH2|ECYT3|HIF-prolyl hydroxylase 2|PHD2SM20|zinc finger MYND domain-containing protein 6|SM-20|EC 1.14.11
- Storage Buffer:50mM Sodium Borate
- Storage Temperature:Store at 4C in the dark.
- Immunogen:The epitope recognized by this antibody maps to a region between residues 1 and 50 of human PHD2/HIF Prolyl Hydroxylase 2 using the numbering given in entry NP_071334.1 (GeneID 54583).
- Purification:Immunogen affinity purified
- Cat. No.:103368-180
Specifications
About this item
The EGLN1 / PHD2 Antibody [DyLight 405] from Novus Biologicals is a rabbit polyclonal antibody to EGLN1 / PHD2. This antibody reacts with human, mouse, rat, primate. The EGLN1 / PHD2 Antibody [DyLight 405] has been validated for the following applications: Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry / Immunofluorescence, Immunohistochemistry-Paraffin.
Type: Primary
Antigen: EGLN1/PHD2
Clonality: Polyclonal
Clone:
Conjugation: DyLight 405
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat, Primate