Order Entry
United States
ContactUsLinkComponent
Anti-EAAT1/GLAST-1/SLC1A3 Rabbit Polyclonal Antibody
Anti-EAAT1/GLAST-1/SLC1A3 Rabbit Polyclonal Antibody
Catalog # 103368-752
Supplier:  Novus Biologicals
Anti-EAAT1/GLAST-1/SLC1A3 Rabbit Polyclonal Antibody
Catalog # 103368-752
Supplier:  Novus Biologicals
Supplier Number:  NB100-1869SS

Specifications

  • Antibody Type:
    Primary
  • Antigen Symbol:
    EAAT1/GLAST-1/SLC1A3
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Flow Cytometry:
    Yes
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • ImmunoFluorescence:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Western Blot:
    Yes
  • Size:
    0.025 ml
  • Gene ID:
    6507
  • Antigen Synonyms:
    GLASTGLAST1|solute carrier family 1 (glial high affinity glutamate transporter)|GLAST|Solute carrier family 1 member 3|EAAT1GLAST-1|member 3|EA6FLJ25094|Sodium-dependent glutamate/aspartate transporter 1|excitatory amino acid transporter 1
  • Storage Buffer:
    PBS
  • Storage Temperature:
    Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
  • Concentration:
    1.0 mg/ml
  • Immunogen:
    A synthetic peptide made to a C-terminal portion of the rat SLC1A3 protein (between residues 500-542) [UniProt P24942]
  • Purification:
    Immunogen affinity purified
  • Cat. No.:
    103368-752

Specifications

About this item

The EAAT1 / GLAST-1 / SLC1A3 Antibody from Novus Biologicals is a rabbit polyclonal antibody to EAAT1 / GLAST-1 / SLC1A3. This antibody reacts with human, mouse, rat. The EAAT1 / GLAST-1 / SLC1A3 Antibody has been validated for the following applications: Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry / Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, Flow (Intracellular).

Type: Primary
Antigen: EAAT1/GLAST-1/SLC1A3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat