Anti-DYNC1LI1 Rabbit Polyclonal Antibody
Catalog # 103274-448
Supplier: Novus Biologicals
Specifications
- Antibody Type:Primary
- Antigen Symbol:DYNC1LI1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human
- Western Blot:Yes
- Size:100 μl
- Gene ID:51143
- Antigen Synonyms:cytoplasmic 1|DNCLI1|cytoplasmic dynein 1 light intermediate chain 1|dynein|light intermediate chain 1|LIC1
- Storage Buffer:PBS (pH 7.2) and 40% Glycerol
- Storage Temperature:Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
- Immunogen:This antibody was developed against Recombinant Protein corresponding to amino acids:AGGGSSGLPPSTKKSGQKPVLDVHAELDRITRKPVTVSPTTPTSPTEGEAS
- Purification:Immunogen affinity purified
- Cat. No.:103274-448
- Supplier no.:NBP1-87972
Specifications
About this item
The DYNC1LI1 Antibody from Novus Biologicals is a rabbit polyclonal antibody to DYNC1LI1. This antibody reacts with human. The DYNC1LI1 Antibody has been validated for the following applications: Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
Type: Primary
Antigen: DYNC1LI1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human