Anti-DRAK2 Mouse Monoclonal Antibody [clone: 2C3]
103336-574
- Antibody Type:Primary
- Antigen Symbol:DRAK2
- Clonality:Monoclonal
- Clone:2C3
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG2b kappa
- Reactivity:Human
- Western Blot:Yes
- Size:100 μg
- Gene ID:9262
- Antigen Synonyms:EC 2.7.11.1|serine/threonine-protein kinase 17B|DRAK2death-associated protein kinase-related 2|DAP kinase-related apoptosis-inducing protein kinase 2|serine/threonine kinase 17b|EC 2.7.11|serine/threonine kinase 17b (apoptosis-inducing)
- Storage Buffer:PBS (pH 7.4)
- Storage Temperature:Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
- Immunogen:STK17B (AAH16040, 272 a.a. ~ 372 a.a) partial recombinant protein with GST tag.SLLVKNPEKRPTAEICLSHSWLQQWDFENLFHPEETSSSSQTQDHSVRSSEDKTSKSSCNGTCGDREDKENIPEDSSMVSKRFRFDDSLPNPHELVSDLLC
- Purification:IgG purified
- Cat. No.:103336-574
- Supplier no.:H00009262-M01
The DRAK2 Antibody (2C3) from Novus Biologicals is a mouse monoclonal antibody to DRAK2. This antibody reacts with human. The DRAK2 Antibody (2C3) has been validated for the following applications: Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry / Immunofluorescence, Immunohistochemistry-Paraffin.
Type: Primary
Antigen: DRAK2
Clonality: Monoclonal
Clone: 2C3
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b Kappa
Reactivity: Human