Order Entry
United States
ContactUsLinkComponent
Anti-DNMT3A Mouse Monoclonal Antibody (DyLight 405) [clone: 64B814.1]
Anti-DNMT3A Mouse Monoclonal Antibody (DyLight 405) [clone: 64B814.1]
Catalog # 103359-648
Supplier:  Novus Biologicals
Anti-DNMT3A Mouse Monoclonal Antibody (DyLight 405) [clone: 64B814.1]
Catalog # 103359-648
Supplier:  Novus Biologicals
Supplier Number:  NB100-56521V
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Antibody Type:
    Primary
  • Antigen Symbol:
    DNMT3A
  • Clonality:
    Monoclonal
  • Clone:
    64B814.1
  • Conjugation:
    DyLight® 405
  • Flow Cytometry:
    Yes
  • Host:
    Mouse
  • ImmunoFluorescence:
    Yes
  • Isotype:
    IgG1 kappa
  • Reactivity:
    Human,
    Mouse
  • Western Blot:
    Yes
  • Size:
    100 μl
  • Environmentally Preferable:
  • Gene ID:
    1788
  • Antigen Synonyms:
    DNA cytosine methyltransferase 3A2|DNA (cytosine-5-)-methyltransferase 3 alpha|DNMT3A2|DNA (cytosine-5)-methyltransferase 3A|DNA MTase HsaIIIA|M.HsaIIIA|EC 2.1.1.37|Dnmt3a|DNA methyltransferase HsaIIIA
  • Storage Buffer:
    50mM Sodium Borate
  • Storage Temperature:
    Store at 4C in the dark.
  • Immunogen:
    This antibody was raised against bacteria expressed HIS-tag recombinant mouse Dnmt3a.
  • Purification:
    Protein G purified
  • Cat. No.:
    103359-648

Specifications

About this item

The DNMT3A Antibody (64B814.1) [DyLight 405] from Novus Biologicals is a mouse monoclonal antibody to DNMT3A. This antibody reacts with human, mouse. The DNMT3A Antibody (64B814.1) [DyLight 405] has been validated for the following applications: Western Blot, Immunocytochemistry / Immunofluorescence, Flow (Intracellular).

Type: Primary
Antigen: DNMT3A
Clonality: Monoclonal
Clone: 64B814.1
Conjugation: DyLight 405
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human, Mouse