Anti-DMP-1 Mouse Monoclonal Antibody [clone: 1D4]
Catalog # 103325-204
Supplier: Novus Biologicals
Specifications
- Antibody Type:Primary
- Antigen Symbol:DMP-1
- Clonality:Monoclonal
- Clone:1D4
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- ImmunoFluorescence:Yes
- Isotype:IgG2b kappa
- Reactivity:Human
- Western Blot:Yes
- Size:100 μg
- Gene ID:1758
- Antigen Synonyms:Dentin matrix protein 1|DMP-1|dentin matrix acidic phosphoprotein 1|ARHP|dentin matrix acidic phosphoprotein|ARHR
- Storage Buffer:PBS (pH 7.4)
- Storage Temperature:Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
- Immunogen:DMP1 (NP_004398.1, 221 a.a. - 330 a.a.) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. ESIRSERGNSRMNSAGMKSKESGENSEQANTQDSGGSQLLEHPSRKIFRKSRISEEDDRSELDDNNTMEEVKSDSTENSNSRDTGLSQPRRDSKGDSQEDSKENLSQEES
- Purification:IgG purified
- Cat. No.:103325-204
- Supplier no.:H00001758-M01
Specifications
About this item
The DMP-1 Antibody (1D4) from Novus Biologicals is a mouse monoclonal antibody to DMP-1. This antibody reacts with human. The DMP-1 Antibody (1D4) has been validated for the following applications: Western Blot, ELISA, Immunocytochemistry / Immunofluorescence.
Type: Primary
Antigen: DMP-1
Clonality: Monoclonal
Clone: 1D4
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b Kappa
Reactivity: Human