Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specifications
- Antibody Type:Primary
- Antigen Symbol:DEP-1/CD148
- Clonality:Monoclonal
- Clone:MEM-CD148/05
- Conjugation:Unconjugated
- Flow Cytometry:Yes
- Host:Mouse
- Isotype:IgG2b
- Reactivity:Human
- Size:100 μg
- Gene ID:5795
- Antigen Synonyms:human density enhanced phosphatase-1|SCC1|mouse|DEP-1|Protein-tyrosine phosphatase receptor type J|receptor type|Density-enhanced phosphatase 1|receptor-type tyrosine-protein phosphatase eta|CD148|Protein-tyrosine phosphatase eta|R-PTP-eta|DEP1EC 3.1.3.48|J polypeptide|protein tyrosine phosphatase|HPTP eta|homolog of|R-PTP-J|CD148 antigen|HPTPeta|susceptibility to colon cancer 1|J
- Storage Buffer:PBS (pH 7.4)
- Storage Temperature:Store at 4C. Do not freeze.
- Concentration:1.0 mg/ml
- Immunogen:Human recombinant CD148 (amino acids 1-444).
- Purification:Protein A purified
- Cat. No.:102713-620
- Supplier no.:NBP2-21790
Specifications
About this item
The DEP-1 / CD148 Antibody (MEM-CD148 / 05) from Novus Biologicals is a mouse monoclonal antibody to DEP-1 / CD148. This antibody reacts with human. The DEP-1 / CD148 Antibody (MEM-CD148 / 05) has been validated for the following applications: Flow Cytometry, CyTOF-ready.
Type: Primary
Antigen: DEP-1/CD148
Clonality: Monoclonal
Clone: MEM-CD148/05
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human