Anti-DEFB118 Rabbit Polyclonal Antibody
Catalog # 103280-554
Supplier: Novus Biologicals
Specifications
- Antibody Type:Primary
- Antigen Symbol:DEFB118
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human
- Size:100 μl
- Gene ID:117285
- Antigen Synonyms:ESP13.6|beta-defensin 118|beta 118|ESC42|DEFB-18|DEFB18|beta 18|C20orf63|defensin
- Storage Buffer:PBS (pH 7.2) and 40% Glycerol
- Storage Temperature:Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
- Immunogen:This antibody was developed against Recombinant Protein corresponding to amino acids:KCWNRSGHCRKQCKDGEAVKDTCKNLRACCIPSNEDHRRVPATSPTPLSDSTPGIIDDILTVRFTTDYFEVSSKKDMVEESEAGRGTETSLPNVHHSS
- Purification:Immunogen affinity purified
- Cat. No.:103280-554
- Supplier no.:NBP1-90695
Specifications
About this item
The DEFB118 Antibody from Novus Biologicals is a rabbit polyclonal antibody to DEFB118. This antibody reacts with human. The DEFB118 Antibody has been validated for the following applications: Immunohistochemistry, Immunohistochemistry-Paraffin.
Type: Primary
Antigen: DEFB118
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human