Anti-DDIT4L Rabbit Polyclonal Antibody
Catalog # 103346-278
Supplier: Novus Biologicals
Specifications
- Antibody Type:Primary
- Antigen Symbol:DDIT4L
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Western Blot:Yes
- Size:100 μg
- Gene ID:115265
- Antigen Synonyms:REDD2Rtp801L|regulated in development and DNA damage response 2|REDD-2|RTP801L|homolog of mouse SMHS1|HIF-1 responsive protein RTP801-like|Protein regulated in development and DNA damage response 2|DNA-damage-inducible transcript 4-like|DNA damage-inducible transcript 4-like protein
- Storage Buffer:PBS (pH 7.4)
- Storage Temperature:Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
- Immunogen:DDIT4L (NP_660287.1, 1 a.a. ~ 193 a.a) full-length human protein.MVATGSLSSKNPASISELLDCGYHPESLLSDFDYWDYVVPEPNLNEVIFEESTCQNLVKMLENCLSKSKQTKLGCSKVLVPEKLTQRIAQDVLRLSSTEPCGLRGCVMHVNLEIENVCKKLDRIVCDSSVVPTFELTLVFKQENCSWTSFRDFFFSRGRFSSGFRRTLILSSGFRLVKKKLYSLIGTTVIEGS
- Purification:Protein G purified
- Cat. No.:103346-278
- Supplier no.:H00115265-D01P
Specifications
About this item
The DDIT4L Antibody from Novus Biologicals is a rabbit polyclonal antibody to DDIT4L. This antibody reacts with human. The DDIT4L Antibody has been validated for the following applications: Western Blot, ELISA.
Type: Primary
Antigen: DDIT4L
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human