To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
- Antibody Type:Primary
- Antigen Symbol:DC-SIGNR/CD299/CLEC4M
- Clonality:Monoclonal
- Clone:2G1
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:IgG3 lambda
- Reactivity:Human
- Western Blot:Yes
- Size:100 μg
- Gene ID:10332
- Antigen Synonyms:DCSIGNRMGC47866|Dendritic cell-specific ICAM-3-grabbing non-integrin 2|DC-SIGNRDC-SIGN-related protein|HP10347|CD299|CD209L1|mannose binding C-type lectin DC-SIGNR|LSIGNC-type lectin domain family 4 member M|CD299 antigenMGC129964|Liver/lymph node-specific ICAM-3-grabbing non-integrin|liver/lymph node-specific ICAM-3 grabbing non-integrin|DC-SIGN2CD209 antigen-like protein 1|CD209Ldendritic cell-specific ICAM-3-grabbing nonintegrin 2|L-SIGN|C-type lectin domain family 4|member M
- Storage Buffer:PBS (pH 7.4)
- Storage Temperature:Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
- Immunogen:CLEC4M (NP_055072, 285 a.a. ~ 395 a.a) partial recombinant protein with GST tag.SQRNWHDSVTACQEVRAQLVVIKTAEEQNFLQLQTSRSNRFSWMGLSDLNQEGTWQWVDGSPLSPSFQRYWNSGEPNNSGNEDCAEFSGSGWNDNRCDVDNYWICKKPAA
- Purification:IgG purified
- Cat. No.:103338-672
- Supplier no.:H00010332-M01
The DC-SIGNR / CD299 / CLEC4M Antibody (2G1) from Novus Biologicals is a mouse monoclonal antibody to DC-SIGNR / CD299 / CLEC4M. This antibody reacts with human. The DC-SIGNR / CD299 / CLEC4M Antibody (2G1) has been validated for the following applications: Western Blot, ELISA.
Type: Primary
Antigen: DC-SIGNR/CD299/CLEC4M
Clonality: Monoclonal
Clone: 2G1
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG3 Lambda
Reactivity: Human