To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Antibody Type:Primary
- Antigen Symbol:DC-SIGN/CD209
- Clonality:Monoclonal
- Clone:109H12.03
- Conjugation:Unconjugated
- Flow Cytometry:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG2a
- Reactivity:Human
- Size:100 μg
- Environmentally Preferable:
- Gene ID:30835
- Antigen Synonyms:CD209 antigendendritic cell-specific intracellular adhesion molecules (ICAM)-3 grabbingnon-integrin|CLEC4LC-type lectin domain family 4 member L|DC-SIGNMGC129965|member L|Dendritic cell-specific ICAM-3-grabbing non-integrin 1|DC-SIGN1C-type lectin domain family 4|CD209 molecule|CDSIGNHIV gpl20-binding protein
- Storage Buffer:Tris-NaCl (pH 8.0)
- Storage Temperature:Store at -20C in the dark. Avoid freeze-thaw cycles.
- Immunogen:human DC-SIGN-transfected HeLa cells
- Purification:Ion exchange chromatography
- Cat. No.:103318-772
- Supplier no.:DDX0207
Specifications
About this item
The DC-SIGN / CD209 Antibody (109H12.03) from Novus Biologicals is a mouse monoclonal antibody to DC-SIGN / CD209. This antibody reacts with human. The DC-SIGN / CD209 Antibody (109H12.03) has been validated for the following applications: Flow Cytometry, Immunohistochemistry, Immunocytochemistry / Immunofluorescence, Immunohistochemistry-Paraffin.
Type: Primary
Antigen: DC-SIGN/CD209
Clonality: Monoclonal
Clone: 109H12.03
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human