Anti-DC-LAMP Mouse Monoclonal Antibody (Biotin) [clone: 109G3]
Catalog # 103318-744
Supplier: Novus Biologicals
Specifications
- Antibody Type:Primary
- Antigen Symbol:DC-LAMP
- Clonality:Monoclonal
- Clone:109G3
- Conjugation:Biotin
- Flow Cytometry:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgG1
- Reactivity:Human
- Size:100 μg
- Gene ID:27074
- Antigen Synonyms:TSC403CD208|lysosome-associated membrane glycoprotein 3|CD208 antigen|LAMP|LAMP-3|lysosomal-associated membrane protein 3DC LAMP|DC-LAMP|Protein TSC403|DCLAMPDC-lysosome-associated membrane glycoprotein
- Storage Buffer:PBS and 50% Glycerol
- Storage Temperature:Store at -20C. Avoid freeze-thaw cycles.
- Immunogen:DC-LAMP/CD208 was used as immunogen.
- Purification:Ion exchange chromatography
- Cat. No.:103318-744
- Supplier no.:DDX0193B-100
Specifications
About this item
The DC-LAMP Antibody (109G3) [Biotin] from Novus Biologicals is a mouse monoclonal antibody to DC-LAMP. This antibody reacts with human. The DC-LAMP Antibody (109G3) [Biotin] has been validated for the following applications: Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Frozen.
Type: Primary
Antigen: DC-LAMP
Clonality: Monoclonal
Clone: 109G3
Conjugation: Biotin
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human