Anti-DAZAP2 Mouse Monoclonal Antibody [clone: 3G21]
Catalog # 103337-624
Supplier: Novus Biologicals
Specifications
- Antibody Type:Primary
- Antigen Symbol:DAZAP2
- Clonality:Monoclonal
- Clone:3G21
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- ImmunoFluorescence:Yes
- Isotype:IgG2b kappa
- Reactivity:Human
- Western Blot:Yes
- Size:100 μg
- Gene ID:9802
- Antigen Synonyms:MGC766|proline-rich transcript|DAZ associated protein 2|KIAA0058DAZ-associated protein 2|brain-expressed protein|proline-rich transcript in brain|MGC14319|PRTB|deleted in azoospermia associated protein 2|Deleted in azoospermia-associated protein 2
- Storage Buffer:PBS (pH 7.4)
- Storage Temperature:Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
- Immunogen:DAZAP2 (NP_055579, 93 a.a. ~ 169 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
- Purification:IgG purified
- Cat. No.:103337-624
- Supplier no.:H00009802-M01
Specifications
About this item
The DAZAP2 Antibody (3G21) from Novus Biologicals is a mouse monoclonal antibody to DAZAP2. This antibody reacts with human. The DAZAP2 Antibody (3G21) has been validated for the following applications: Western Blot, ELISA, Immunocytochemistry / Immunofluorescence.
Type: Primary
Antigen: DAZAP2
Clonality: Monoclonal
Clone: 3G21
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b Kappa
Reactivity: Human