Anti-DARS Rabbit Polyclonal Antibody
Catalog # 103273-688
Supplier: Novus Biologicals
Specifications
- Antibody Type:Primary
- Antigen Symbol:DARS
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human
- Western Blot:Yes
- Size:100 μl
- Gene ID:1615
- Antigen Synonyms:Cell proliferation-inducing gene 40 protein|cytoplasmic|MGC111579|EC 6.1.1.12|cell proliferation-inducing protein 40|aspartyl-tRNA synthetase|DKFZp781B11202|aspartate tRNA ligase 1|Aspartate--tRNA ligase|aspRS
- Storage Buffer:PBS (pH 7.2) and 40% Glycerol
- Storage Temperature:Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
- Immunogen:This antibody was developed against Recombinant Protein corresponding to amino acids:LHVQKIYVISLAEPRLPLQLDDAVRPEAEGEEEGRATVNQDTRLDNRVIDLRTSTSQAVFRLQSGICHLFRETLINKGFVEIQTPKIISAASEGGANVFTVSYFKNNAYLAQSPQLYKQMCICAD
- Purification:Immunogen affinity purified
- Cat. No.:103273-688
- Supplier no.:NBP1-86027
Specifications
About this item
The DARS Antibody from Novus Biologicals is a rabbit polyclonal antibody to DARS. This antibody reacts with human. The DARS Antibody has been validated for the following applications: Western Blot, Immunohistochemistry, Immunocytochemistry / Immunofluorescence, Immunohistochemistry-Paraffin.
Type: Primary
Antigen: DARS
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human