Anti-D4S234E Mouse Monoclonal Antibody [clone: 1C3]
103342-518
- Antibody Type:Primary
- Antigen Symbol:D4S234E
- Clonality:Monoclonal
- Clone:1C3
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:IgG1 kappa
- Reactivity:Human,Rat
- Western Blot:Yes
- Size:100 μg
- Gene ID:27065
- Antigen Synonyms:NSG1|DNA segment on chromosome 4 (unique) 234 expressed sequence|P21|D4S234Brain neuron cytoplasmic protein 1|carboxyterminally EE-tagged neuron-enriched endosomal 21 kDa protein|NEEP21|neuron-specific protein family member 1
- Storage Buffer:PBS (pH 7.4)
- Storage Temperature:Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
- Immunogen:D4S234E (AAH01745, 1 a.a. ~ 186 a.a) full length recombinant protein with GST tag.MVKLGNNFAEKGTKQPLLEDGFDTIPLMTPLDVNQLQFPPPDKVVVKTKTEYEPDRKKGKARPPQIAEFTVSITEGVTERFKVSVLVLFALAFLTCVVFLVVYKVYKYDRACPDGFVLKNTQCIPEGLESYYAEQDSSAREKFYTVINHYNLAKQSITRSVSPWMSVLSEEKLSEQETEAAEKSA
- Purification:IgG purified
- Cat. No.:103342-518
- Supplier no.:H00027065-M01
The D4S234E Antibody (1C3) from Novus Biologicals is a mouse monoclonal antibody to D4S234E. This antibody reacts with human, rat. The D4S234E Antibody (1C3) has been validated for the following applications: Western Blot, ELISA.
Type: Primary
Antigen: D4S234E
Clonality: Monoclonal
Clone: 1C3
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human, Rat