Anti-CYP4F3 Mouse Monoclonal Antibody [clone: 4E11.]
Catalog # 103330-248
Supplier: Novus Biologicals
Specifications
- Antibody Type:Primary
- Antigen Symbol:CYP4F3
- Clonality:Monoclonal
- Clone:4E11.
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:IgG2b kappa
- Reactivity:Human
- Western Blot:Yes
- Size:100 μg
- Gene ID:4051
- Antigen Synonyms:polypeptide 3|leukotriene-B4 20-monooxygenase|Leukotriene-B(4) 20-monooxygenase 2|subfamily IVF|EC 1.14.13.30|cytochrome P-450|cytochrome P450|CYP4F|polypeptide 3 (leukotriene B4 omegahydroxylase)|CYPIVF3|subfamily F|LTB4HCPF3|leukotriene-B(4) omega-hydroxylase 2|Cytochrome P450 4F3|leukotriene B4 omega hydroxylase|family 4|Cytochrome P450-LTB-omega
- Storage Buffer:PBS (pH 7.4)
- Storage Temperature:Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
- Immunogen:CYP4F3 (NP_000887, 100 a.a. - 198 a.a.) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. RIFHPTYIKPVLFAPAAIVPKDKVFYSFLKPWLGDGLLLSAGEKWSRHRRMLTPAFHFNILKPYMKIFNESVNIMHAKWQLLASEGSARLDMFEHISLM
- Purification:IgG purified
- Cat. No.:103330-248
- Supplier no.:H00004051-M01
Specifications
About this item
The CYP4F3 Antibody (4E11.) from Novus Biologicals is a mouse monoclonal antibody to CYP4F3. This antibody reacts with human. The CYP4F3 Antibody (4E11.) has been validated for the following applications: Western Blot, ELISA.
Type: Primary
Antigen: CYP4F3
Clonality: Monoclonal
Clone: 4E11.
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b Kappa
Reactivity: Human