Anti-CYFIP2 Mouse Monoclonal Antibody [clone: 4G6]
Catalog # 103342-472
Supplier: Novus Biologicals
Specifications
- Antibody Type:Primary
- Antigen Symbol:CYFIP2
- Clonality:Monoclonal
- Clone:4G6
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:IgG2a kappa
- Reactivity:Human
- Western Blot:Yes
- Size:100 μg
- Gene ID:26999
- Antigen Synonyms:KIAA1168|cytoplasmic FMR1 interacting protein 2|PIR121p53-inducible protein 121|cytoplasmic FMR1-interacting protein 2|p53 inducible protein
- Storage Buffer:PBS (pH 7.4)
- Storage Temperature:Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
- Immunogen:CYFIP2 (NP_055191, 733 a.a. ~ 821 a.a) partial recombinant protein with GST tag.YGVIIPYPPSNRYETLLKQRHVQLLGRSIDLNRLITQRISAAMYKSLDQAISRFESEDLTSIVELEWLLEINRLTHRLLCKHMTLDSF
- Purification:IgG purified
- Cat. No.:103342-472
- Supplier no.:H00026999-M01
Specifications
About this item
The CYFIP2 Antibody (4G6) from Novus Biologicals is a mouse monoclonal antibody to CYFIP2. This antibody reacts with human. The CYFIP2 Antibody (4G6) has been validated for the following applications: Western Blot, ELISA.
Type: Primary
Antigen: CYFIP2
Clonality: Monoclonal
Clone: 4G6
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a Kappa
Reactivity: Human