Anti-CX3CR1 Mouse Polyclonal Antibody
Catalog # 103324-704
Supplier: Novus Biologicals
Specifications
- Antibody Type:Primary
- Antigen Symbol:CX3CR1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human
- Size:0.05 mg
- Gene ID:1524
- Antigen Synonyms:GPR13G-protein coupled receptor 13|Beta chemokine receptor-like 1|CMKBRL1|CCRL1|G protein-coupled receptor 13|CMKDR1|CX3C chemokine receptor 1|Fractalkine receptor|CMK-BRL1|chemokine (C-C) receptor-like 1|V28C-X3-C CKR-1|chemokine (C-X3-C motif) receptor 1|chemokine (C-X3-C) receptor 1|CMK-BRL-1|GPRV28
- Storage Buffer:PBS (pH 7.4)
- Storage Temperature:Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
- Immunogen:CX3CR1 (NP_001328.1, 1 a.a. - 355 a.a.) full-length human protein. MDQFPESVTENFEYDDLAEACYIGDIVVFGTVFLSIFYSVIFAIGLVGNLLVVFALTNSKKPKSVTDIYLLNLALSDLLFVATLPFWTHYLINEKGLHNAMCKFTTAFFFIGFFGSIFFITVISIDRYLAIVLAANSMNNRTVQHGVTISLGVWAAAILVAAPQFMFTKQKENECLGDYPEVLQEIWPVLRNVETNFLGFLLPLLIMSYCYFRIIQTLFSCKNHKKAKAIKLILLVVIVFFLFWTPYNVMIFLETLKLYDFFPSCDMRKDLRLALSVTETVAFSHCCLNPLIYAFAGEKFRRYLYHLYGKCLAVLCGRSVHVDFSSSESQRSRHGSVLSSNFTYHTSDGDALLLL
- Purification:Protein A purified
- Cat. No.:103324-704
- Supplier no.:H00001524-B01P
Specifications
About this item
The CX3CR1 Antibody from Novus Biologicals is a mouse polyclonal antibody to CX3CR1. This antibody reacts with human. The CX3CR1 Antibody has been validated for the following applications: Immunocytochemistry / Immunofluorescence.
Type: Primary
Antigen: CX3CR1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human