Anti-CTAGE5 Rabbit Polyclonal Antibody
Catalog # 103271-986
Supplier: Novus Biologicals
Specifications
- Antibody Type:Primary
- Antigen Symbol:CTAGE5
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human
- Western Blot:Yes
- Size:100 μl
- Gene ID:4253
- Antigen Synonyms:MGEA|meningioma expressed antigen 6 (coiled-coil proline-rich)|MEA11|cTAGE-5A|cTAGE-5D|MGEA6|MGEA11cTAGE-5C|cutaneous T-cell lymphoma-associated antigen 5|member 5|cTAGE family member 5|Protein cTAGE-5|Meningioma-expressed antigen 6/11|CTAGE family|MEA6cTAGE-5B
- Storage Buffer:PBS (pH 7.2) and 40% Glycerol
- Storage Temperature:Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
- Immunogen:This antibody was developed against Recombinant Protein corresponding to amino acids:EKEATEAQSLEATCEKLNRSNSELEDEILCLEKELKEEKSKHSEQDELMADISKRIQSLEDESKSLKSQVAEAKMTFKIFQMNEERLKIAIKDALNENSQLQESQKQLLQ
- Purification:Immunogen affinity purified
- Cat. No.:103271-986
- Supplier no.:NBP1-85390
Specifications
About this item
The CTAGE5 Antibody from Novus Biologicals is a rabbit polyclonal antibody to CTAGE5. This antibody reacts with human. The CTAGE5 Antibody has been validated for the following applications: Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
Type: Primary
Antigen: CTAGE5
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human