Anti-CRTAP Rabbit Polyclonal Antibody
Catalog # 103285-376
Supplier: Novus Biologicals
Specifications
- Antibody Type:Primary
- Antigen Symbol:CRTAP
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human
- Western Blot:Yes
- Size:100 μl
- Gene ID:10491
- Antigen Synonyms:cartilage-associated protein|CASPLEPREL3|cartilage associated protein|leprecan-like 3
- Storage Buffer:PBS (pH 7.2) and 40% Glycerol
- Storage Temperature:Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
- Immunogen:This antibody was developed against Recombinant Protein corresponding to amino acids:RAHCLKRCKQGLPAFRQSQPSREVLADFQRREPYKFLQFAYFKANNLPKAIAAAHTFLLKHPDDEMMKRNMAYYKSLPGAEDYIKDLE
- Purification:Immunogen affinity purified
- Cat. No.:103285-376
- Supplier no.:NBP1-94104
Specifications
About this item
The CRTAP Antibody from Novus Biologicals is a rabbit polyclonal antibody to CRTAP. This antibody reacts with human. The CRTAP Antibody has been validated for the following applications: Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
Type: Primary
Antigen: CRTAP
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human