Anti-CLK3 Mouse Monoclonal Antibody [clone: 1H2]
Catalog # 103324-070
Supplier: Novus Biologicals
Specifications
- Antibody Type:Primary
- Antigen Symbol:CLK3
- Clonality:Monoclonal
- Clone:1H2
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgG2b kappa
- Reactivity:Human,Rat
- Western Blot:Yes
- Size:100 μg
- Gene ID:1198
- Antigen Synonyms:PHCLK3/152|FLJ22858|dual specificity protein kinase CLK3|CDC-like kinase 3PHCLK3|EC 2.7.12.1|clk3
- Storage Buffer:PBS (pH 7.4)
- Storage Temperature:Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
- Immunogen:CLK3 (AAH02555, 36 a.a. - 136 a.a.) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. YPSRREPPPRRSRSRSHDRLPYQRRYRERRDSDTYRCEERSPSFGEDYYGPSRSRHRRRSRERGPYRTRKHAHHCHKRRTRSCSSASSRSQQSSKRSSRSV
- Purification:IgG purified
- Cat. No.:103324-070
- Supplier no.:H00001198-M01
Specifications
About this item
The CLK3 Antibody (1H2) from Novus Biologicals is a mouse monoclonal antibody to CLK3. This antibody reacts with human, rat. The CLK3 Antibody (1H2) has been validated for the following applications: Western Blot, ELISA, Immunohistochemistry-Paraffin.
Type: Primary
Antigen: CLK3
Clonality: Monoclonal
Clone: 1H2
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b Kappa
Reactivity: Human, Rat