Anti-CHRND Rabbit Polyclonal Antibody
103323-968
: Avantor
- Antibody Type:Primary
- Antigen Symbol:CHRND
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Western Blot:Yes
- Size:100 μg
- Gene ID:1144
- Antigen Synonyms:CMS2A|nicotinic|acetylcholine receptor subunit delta|delta polypeptide|delta|cholinergic receptor|FCCMS|ACHRDSCCMS
- Storage Buffer:PBS (pH 7.4)
- Storage Temperature:Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
- Immunogen:CHRND (NP_000742.1, 1 a.a. - 517 a.a.) full-length human protein. MEGPVLTLGLLAALAVCGSWGLNEEERLIRHLFQEKGYNKELRPVAHKEESVDVALALTLSNLISLKEVEETLTTNVWIEHGWTDNRLKWNAEEFGNISVLRLPPDMVWLPEIVLENNNDGSFQISYSCNVLVYHYGFVYWLPPAIFRSSCPISVTYFPFDWQNCSLKFSSLKYTAKEITLSLKQDAKENRTYPVEWIIIDPEGFTENGEWEIVHRPARVNVDPRAPLDSPSRQDITFYLIIRRKPLFYIINILVPCVLISFMVNLVFYLPADSGEKTSVAISVLLAQSVFLLLISKRLPATSMAIPLIGKFLLFGMVLVTMVVVICVIVLNIHFRTPSTHVLSEGVKKLFLETLPELLHMSRPAEDGPSPGALVRRSSSLGYISKAEEYFLLKSRSDLMFEKQSERHGLARRLTTARRPPASSEQAQQELFNELKPAVDGANFIVNHMRDQNNYNEEKDSWNRVARTVDRLCLFVVTPVMVVGTAWIFLQGVYNQPPPQPFPGDPYSYNVQDKRFI
- Purification:Protein A purified
- Cat. No.:103323-968
- Supplier no.:H00001144-D01P
The CHRND Antibody from Novus Biologicals is a rabbit polyclonal antibody to CHRND. This antibody reacts with human. The CHRND Antibody has been validated for the following applications: Western Blot.
Type: Primary
Antigen: CHRND
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human