Anti-CGR19 Mouse Monoclonal Antibody [clone: 2E12]
Catalog # 103339-412
Supplier: Novus Biologicals
Specifications
- Antibody Type:Primary
- Antigen Symbol:CGR19
- Clonality:Monoclonal
- Clone:2E12
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:IgG1 kappa
- Reactivity:Human
- Size:100 μg
- Gene ID:10668
- Antigen Synonyms:RING finger protein 197|Cell growth regulatory gene 19 protein|cell growth regulator with ring finger domain 1|CGR19cell growth regulator with RING finger domain protein 1|RNF197GCRRF1
- Storage Buffer:PBS (pH 7.4)
- Storage Temperature:Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
- Immunogen:CGRRF1 (AAH15063, 36 a.a. ~ 333 a.a) full length recombinant protein with GST tag.MAAVFLVTLYEYSPLFYIAVVFTCFIVTTGLVLGWFGWDVPVILRNSEETQFSTRVFKKQMRQVKNPFGLEITNPSSASITTGITLTTDCLEDSLLTCYWGCNVQKLYEALQKHVYCFRISTPQALEDALYSEYLYQEQYFIKKDSKEEIYCQLPRDTKIEDFGTVPRSRYPLVALLTLADEDDREIYDIISMVSVIHIPDRTYKLSCRILYQYLLLAQGQFHDLKQLFMSANNNFTPSNNSSSEEKNTDRSLLE
- Purification:IgG purified
- Cat. No.:103339-412
- Supplier no.:H00010668-M01
Specifications
About this item
The CGR19 Antibody (2E12) from Novus Biologicals is a mouse monoclonal antibody to CGR19. This antibody reacts with human. The CGR19 Antibody (2E12) has been validated for the following applications: ELISA.
Type: Primary
Antigen: CGR19
Clonality: Monoclonal
Clone: 2E12
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human