Anti-CEP57 Mouse Polyclonal Antibody
Catalog # 103337-494
Supplier: Novus Biologicals
Specifications
- Antibody Type:Primary
- Antigen Symbol:CEP57
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human
- Western Blot:Yes
- Size:0.05 mg
- Gene ID:9702
- Antigen Synonyms:KIAA0092centrosomal protein of 57 kDa|TSP57proliferation-inducing protein 8|PIG8|Translokin|centrosomal protein 57kDa|Cep57|FGF2-interacting protein|Testis-specific protein 57
- Storage Buffer:PBS (pH 7.4)
- Storage Temperature:Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
- Immunogen:CEP57 (NP_055494.2, 1 a.a. ~ 500 a.a) full-length human protein.MAAASVSAASGSHLSNSFAEPSRSNGSMVRHSSSPYVVYPSDKPFLNSDLRRSPSKPTLAYPESNSRAIFSALKNLQDKIRRLELERIQAEESVKTLSRETIEYKKVLDEQIQERENSKNEESKHNQELTSQLLAAENKCNLLEKQLEYMRNMIKHAEMERTSVLEKQVSLERERQHDQTHVQSQLEKLDLLEQEYNKLTTMQALAEKKMQELEAKLHEEEQERKRMQAKAAELQTGLETNRLIFEDKATPCVPN
- Purification:Protein A purified
- Cat. No.:103337-494
- Supplier no.:H00009702-B01P
Specifications
About this item
The CEP57 Antibody from Novus Biologicals is a mouse polyclonal antibody to CEP57. This antibody reacts with human. The CEP57 Antibody has been validated for the following applications: Western Blot, ELISA, Immunocytochemistry / Immunofluorescence, Immunofluorescence.
Type: Primary
Antigen: CEP57
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human