Anti-CDP/CUTL1 Mouse Monoclonal Antibody [clone: 2D10]
103324-702
- Antibody Type:Primary
- Antigen Symbol:CDP/CUTL1
- Clonality:Monoclonal
- Clone:2D10
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:IgG1 kappa
- Reactivity:Human
- Western Blot:Yes
- Size:100 μg
- Gene ID:1523
- Antigen Synonyms:protein CASP|CUT|p75|CDP/Cux|p200|cut-like 1|cut-like homeobox 1|p100|GOLIM6|cut (Drosophila)-like 1 (CCAAT displacement protein)|putative protein product of Nbla10317|p110|Nbla10317|CCAAT displacement protein (Drosophila)|CDPCCAAT displacement protein|Homeobox protein cux-1|COY1|homeobox protein cut-like 1|FLJ31745|golgi integral membrane protein 6|CCAAT displacement protein
- Storage Buffer:PBS (pH 7.4)
- Storage Temperature:Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
- Immunogen:CUTL1 (NP_001904.2, 521 a.a. - 620 a.a.) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. AENRLAQHTLQALQSELDSLRADNIKLFEKIKFLQSYPGRGSGSDDTELRYSSQYEERLDPFSSFSKRERQRKYLSLSPWDKATLSMGRLVLSNKMARTI
- Purification:IgG purified
- Cat. No.:103324-702
- Supplier no.:H00001523-M02
The CDP / CUTL1 Antibody (2D10) from Novus Biologicals is a mouse monoclonal antibody to CDP / CUTL1. This antibody reacts with human. The CDP / CUTL1 Antibody (2D10) has been validated for the following applications: Western Blot, ELISA.
Type: Primary
Antigen: CDP/CUTL1
Clonality: Monoclonal
Clone: 2D10
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human