Anti-CCNM3 Mouse Polyclonal Antibody
Catalog # 103342-354
Supplier: Novus Biologicals
Specifications
- Antibody Type:Primary
- Antigen Name:Cyclin M3
- Antigen Symbol:CCNM3
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human
- Western Blot:Yes
- Size:0.05 mg
- Gene ID:26505
- Antigen Synonyms:DKFZp434I1016|metal transporter CNNM3|ancient conserved domain protein 3|FLJ20018|cyclin M3|Ancient conserved domain-containing protein 3|Cyclin-M3|ACDP3cyclin-M3
- Storage Buffer:PBS (pH 7.4)
- Storage Temperature:Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
- Immunogen:CNNM3 (AAH07199, 1 a.a. ~ 355 a.a) full-length human protein.MLDASTVLDFGVLASIMQSGHTRIPVYEEERSNIVDMLYLKDLAFVDPEDCTPLSTITRFYNHPLHFVFNDTKLDAVLEEFKRGDTVVKRKPASLMAPLKRKEEFSLFKVSDDEYKVTISPQLLLATQRFLSREVDVFSPLRISEKVLLHLLKHPSVNQEVRFDESNRLATHHYLYQRSQPVDYFILILQGRVEVEIGKEGLKFENGAFTYYGVSALTVPSSVHQSPVSSLQPIRHDLQPDPGDGTHSSAYCPDY
- Purification:Protein A purified
- Cat. No.:103342-354
- Supplier no.:H00026505-B01P
Specifications
About this item
The Cyclin M3 Antibody from Novus Biologicals is a mouse polyclonal antibody to Cyclin M3. This antibody reacts with human. The Cyclin M3 Antibody has been validated for the following applications: Western Blot, ELISA, Immunocytochemistry / Immunofluorescence.
Type: Primary
Antigen: CCNM3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human