SP Bel-Art Acid/Solvent Bottle Carriers, Bel-Art Products, a part of SP
Catalog # 80700-744
Supplier: Bel-Art Products, a Part of SP
Specifications
- Ø:18 cm (7⅛")
- Description:Acid/solvent carrier truncated Lid
- Height:32 cm (12.5")
- Cat. No.:80700-744
- Supplier no.:169590000
Specifications
About this item
Polyethylene carriers with epoxy-coated, heavy-gauge steel wire bails feature wide ribs on both the sides and the bottom to support and cushion bottles.
- Truncated style lid permits pouring without the removal of the lid
- Designed to hold standard 2.5 L (5 pt.) US reagent bottles