Order Entry
United States
ContactUsLinkComponent
1x Trspsin-EDTA Solution from Porcine, MP Biomedicals
1x Trspsin-EDTA Solution from Porcine, MP Biomedicals
Catalog # 76180-672
Supplier:  MP Biomedicals
CAS Number:  
undefined
1x Trspsin-EDTA Solution from Porcine, MP Biomedicals
Catalog # 76180-672
Supplier:  MP Biomedicals
Supplier Number:  091689149
CAS Number:  

Specifications

  • Size:
    100 mL
  • Cat. No.:
    76180-672

Specifications

About this item

One USP trypsin unit is the activity causing a change in absorbance of 0.003 per minute under the conditions specified., Pale to Dark Pink Solution.

  • Sterile
  • Concentration: 0.05% (w/v) Trypsin (1:250) and 0.02% (w/v) EDTA (Versene)
  • Formulation: EDTA 2Na 2H2O: 200.00000 mg/L, Potassium Chloride [KCl]: 400.00000 mg/L, Sodium Bicarbonate [NaHCO3]: 580.00000 mg/L, Sodium Chloride [NaCl]: 8000.00000 mg/L, Dextrose: 1000.00000 mg/L, Phenol Red Sodium Salt: 2.00000 mg/L, Trypsin 1:250: 500.00000 mg/L
  • pH: 7.0-7.4
  • Foreign Activity: Mycoplasma, Porcine Parvovirus : Not detected
  • Osmolality: 270 - 305 mM/kg
  • Storage and Handling: -20 °C

Trypsin-EDTA solution is widely used for cell dissociation, routine cell culture passaging, and primary tissue dissociation due to its digestive strength.