Anti-CBF1-interacting corepressor Mouse Monoclonal Antibody [clone: 2E11]
103337-206
- Antibody Type:Primary
- Antigen Name:CBF1-interacting corepressor
- Clonality:Monoclonal
- Clone:2E11
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:IgG1 kappa
- Reactivity:Human
- Western Blot:Yes
- Size:100 μg
- Gene ID:9541
- Antigen Synonyms:CBF1-interacting corepressor|CIRrecepin|corepressor interacting with RBPJ|Recepin|corepressor interacting with RBPJ 1|CBF1 interacting corepressor|1
- Storage Buffer:PBS (pH 7.4)
- Storage Temperature:Store at -20C. Avoid freeze-thaw cycles.
- Immunogen:CIR (NP_004873, 136 a.a. ~ 205 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.HVNTDRECPLFGLSGINASSVPTDGSGPSMHPSELIAEMRNSGFALKRNVLGRNLTANDPSQEYVASEG
- Purification:Protein A purified
- Cat. No.:103337-206
- Supplier no.:H00009541-M01
The CBF1-interacting corepressor Antibody (2E11) from Novus Biologicals is a mouse monoclonal antibody to CBF1-interacting corepressor. This antibody reacts with human. The CBF1-interacting corepressor Antibody (2E11) has been validated for the following applications: Western Blot, ELISA.
Type: Primary
Antigen: CBF1-interacting corepressor
Clonality: Monoclonal
Clone: 2E11
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human