To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
- Antibody Type:Primary
- Antigen Name:Casein Kinase 1 epsilon
- Clonality:Monoclonal
- Clone:2E1.
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- ImmunoFluorescence:Yes
- Isotype:IgG2a kappa
- Reactivity:Human
- Size:100 μg
- Gene ID:1454
- Antigen Synonyms:MGC10398|epsilon|CKIe|EC 2.7.11.1|casein kinase I isoform epsilon|HCKIE|EC 2.7.11|casein kinase 1|CKI-epsilon
- Storage Buffer:PBS (pH 7.4)
- Storage Temperature:Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
- Immunogen:CSNK1E (NP_689407, 2 a.a. - 100 a.a.) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. ELRVGNKYRLGRKIGSGSFGDIYLGANIASGEEVAIKLECVKTKHPQLHIESKFYKMMQGGVGIPSIKWCGAEGDYNVMVMELLGPSLEDLFNFCSRKF
- Purification:IgG purified
- Cat. No.:103324-508
- Supplier no.:H00001454-M07
The Casein Kinase 1 epsilon Antibody (2E1.) from Novus Biologicals is a mouse monoclonal antibody to Casein Kinase 1 epsilon. This antibody reacts with human. The Casein Kinase 1 epsilon Antibody (2E1.) has been validated for the following applications: ELISA, Immunocytochemistry / Immunofluorescence, Proximity Ligation Assay.
Type: Primary
Antigen: Casein Kinase 1 epsilon
Clonality: Monoclonal
Clone: 2E1.
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a Kappa
Reactivity: Human