Anti-Calcium-sensing R/CaSR Rabbit Polyclonal Antibody
Catalog # 103266-838
Supplier: Novus Biologicals
Specifications
- Antibody Type:Primary
- Antigen Name:Calcium-sensing R/CaSR
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human
- Western Blot:Yes
- Size:100 μl
- Gene ID:846
- Antigen Synonyms:Parathyroid cell calcium-sensing receptor|parathyroid Ca(2+)-sensing receptor 1|EIG8|FHH|PCAR1MGC138441|FIH|hypocalciuric hypercalcemia 1|calcium-sensing receptor|HHC1|GPRC2AHHC|PCaR1|extracellular calcium-sensing receptor|CaSR|CAR|NSHPT
- Storage Buffer:PBS (pH 7.2) and 40% Glycerol
- Storage Temperature:Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
- Immunogen:This antibody was developed against Recombinant Protein corresponding to amino acids:GTVTFSLSFDEPQKNAMAHRNSTHQNSLEAQKSSDTLTRHQPLLPLQCGETDLDLTVQETGLQGPVGGDQRPEVEDPEELSPALVVSSSQSFVISGG
- Purification:Immunogen affinity purified
- Cat. No.:103266-838
- Supplier no.:NBP1-84687
Specifications
About this item
The Calcium-sensing R / CaSR Antibody from Novus Biologicals is a rabbit polyclonal antibody to Calcium-sensing R / CaSR. This antibody reacts with human. The Calcium-sensing R / CaSR Antibody has been validated for the following applications: Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
Type: Primary
Antigen: Calcium-sensing R/CaSR
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human