Order Entry
United States
ContactUsLinkComponent
Anti-Birch pollen Bet V1 Mouse Monoclonal Antibody [clone: 110G10.01]
Anti-Birch pollen Bet V1 Mouse Monoclonal Antibody [clone: 110G10.01]
Catalog # 103318-672
Supplier:  Novus Biologicals
Anti-Birch pollen Bet V1 Mouse Monoclonal Antibody [clone: 110G10.01]
Catalog # 103318-672
Supplier:  Novus Biologicals
Supplier Number:  DDX0051P-100

Specifications

  • Antibody Type:
    Primary
  • Antigen Name:
    Birch pollen Bet V1
  • Clonality:
    Monoclonal
  • Clone:
    110G10.01
  • Conjugation:
    Unconjugated
  • Host:
    Mouse
  • Isotype:
    IgG1
  • Reactivity:
    Plant
  • Western Blot:
    Yes
  • Size:
    100 μg
  • Antigen Synonyms:
    Allergen Bet v I-A|BETVIA|Major pollen allergen Bet v 1-A|Allergen=Bet v 1-A|BETVI
  • Storage Buffer:
    Tris-NaCl (pH 8.0)
  • Storage Temperature:
    Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
  • Concentration:
    0.5 mg/ml
  • Immunogen:
    Birch Pollen
  • Purification:
    Ion exchange chromatography
  • Cat. No.:
    103318-672
  • Supplier no.:
    DDX0051P-100

Specifications

About this item

The Birch pollen Bet V1 Antibody (110G10.01) from Novus Biologicals is a mouse monoclonal antibody to Birch pollen Bet V1. This antibody reacts with plant. The Birch pollen Bet V1 Antibody (110G10.01) has been validated for the following applications: Western Blot.

Type: Primary
Antigen: Birch pollen Bet V1
Clonality: Monoclonal
Clone: 110G10.01
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Plant