To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Antibody Type:Primary
- Antigen Name:Birch pollen Bet V1
- Clonality:Monoclonal
- Clone:H.G17E7A1
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Human
- Isotype:IgG4
- Reactivity:Plant
- Size:100 μg
- Antigen Synonyms:Allergen Bet v I-A|BETVIA|Major pollen allergen Bet v 1-A|Allergen=Bet v 1-A|BETVI
- Storage Buffer:Tris-NaCl (pH 8.0)
- Storage Temperature:Store at -20C. Avoid freeze-thaw cycles.
- Concentration:0.5 mg/ml
- Immunogen:Birch pollen Bet V1 was used as immunogen.
- Purification:Ion exchange chromatography
- Cat. No.:103318-690
- Supplier no.:DDX0076P-100
Specifications
About this item
The Birch pollen Bet V1 Antibody (H.G17E7A1) from Novus Biologicals is a human monoclonal antibody to Birch pollen Bet V1. This antibody reacts with plant. The Birch pollen Bet V1 Antibody (H.G17E7A1) has been validated for the following applications: ELISA, Functional.
Functional
Type: Primary
Antigen: Birch pollen Bet V1
Clonality: Monoclonal
Clone: H.G17E7A1
Conjugation: Unconjugated
Epitope:
Host: Human
Isotype: IgG4
Reactivity: Plant