Order Entry
United States
ContactUsLinkComponent
Anti-beta-Galactosidase-1/GLB1 Mouse Monoclonal Antibody [clone: 6E7.]
Anti-beta-Galactosidase-1/GLB1 Mouse Monoclonal Antibody [clone: 6E7.]
Catalog # 103327-230
Supplier:  Novus Biologicals
Anti-beta-Galactosidase-1/GLB1 Mouse Monoclonal Antibody [clone: 6E7.]
Catalog # 103327-230
Supplier:  Novus Biologicals
Supplier Number:  H00002720-M01

Specifications

  • Antibody Type:
    Primary
  • Antigen Name:
    beta-Galactosidase-1/GLB1
  • Clonality:
    Monoclonal
  • Clone:
    6E7.
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Mouse
  • ImmunoPrecipitation:
    Yes
  • Isotype:
    IgG2a kappa
  • Reactivity:
    Human
  • Western Blot:
    Yes
  • Size:
    100 μg
  • Gene ID:
    2720
  • Antigen Synonyms:
    MPS4B|67kDa|EBP|beta 1|elastin receptor 1 (67kD)|Acid beta-galactosidase|Lactase|Elastin receptor 1|ELNR1|beta-galactosidase|EC 3.2.1.23|galactosidase|elastin receptor 1
  • Storage Buffer:
    PBS (pH 7.4)
  • Storage Temperature:
    Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
  • Immunogen:
    GLB1 (NP_000395 578 a.a. - 677 a.a.) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. KGQVWINGFNLGRYWPARGPQLTLFVPQHILMTSAPNTITVLELEWAPCSSDDPELCAVTFVDRPVIGSSVTYDHPSKPVEKRLMPPPPQKNKDSWLDHV
  • Purification:
    IgG purified
  • Cat. No.:
    103327-230
  • Supplier no.:
    H00002720-M01

Specifications

About this item

The beta-Galactosidase-1 / GLB1 Antibody (6E7.) from Novus Biologicals is a mouse monoclonal antibody to beta-Galactosidase-1 / GLB1. This antibody reacts with human. The beta-Galactosidase-1 / GLB1 Antibody (6E7.) has been validated for the following applications: Western Blot, ELISA, Immunoprecipitation.

Type: Primary
Antigen: beta-Galactosidase-1/GLB1
Clonality: Monoclonal
Clone: 6E7.
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a Kappa
Reactivity: Human