Anti-beta II Tubulin B Mouse Monoclonal Antibody (HRP (Horseradish Peroxidase)) [clone: 5B2]
Catalog # 103402-778
Supplier: Novus Biologicals
Specifications
- Antibody Type:Primary
- Antigen Name:beta II Tubulin B
- Clonality:Monoclonal
- Clone:5B2
- Conjugation:HRP (Horseradish Peroxidase)
- ELISA:Yes
- Host:Mouse
- Isotype:IgG2b kappa
- Reactivity:Human,Mouse
- Western Blot:Yes
- Size:100 μl
- Gene ID:347733
- Antigen Synonyms:DKFZp566F223|bA506K6.1|tubulin|tubulin beta-2B chain|MGC8685|beta 2B|FLJ98847|beta polypeptide paralog|class II beta-tubulin isotype
- Storage Buffer:PBS
- Storage Temperature:Store at 4C in the dark.
- Immunogen:Recombinant human beta-Tubulin (1-445aa) purified from E. coli
- Purification:Protein G purified
- Cat. No.:103402-778
Specifications
About this item
The beta II Tubulin B Antibody (5B2) [HRP] from Novus Biologicals is a mouse monoclonal antibody to beta II Tubulin B. This antibody reacts with human, mouse. The beta II Tubulin B Antibody (5B2) [HRP] has been validated for the following applications: Western Blot, ELISA.
Type: Primary
Antigen: beta II Tubulin B
Clonality: Monoclonal
Clone: 5B2
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Mouse
Isotype: IgG2b Kappa
Reactivity: Human, Mouse