Order Entry
United States
ContactUsLinkComponent
Anti-beta II Tubulin B Mouse Monoclonal Antibody (DyLight 488) [clone: 5B2]
Anti-beta II Tubulin B Mouse Monoclonal Antibody (DyLight 488) [clone: 5B2]
Catalog # 103402-766
Supplier:  Novus Biologicals
Anti-beta II Tubulin B Mouse Monoclonal Antibody (DyLight 488) [clone: 5B2]
Catalog # 103402-766
Supplier:  Novus Biologicals
Supplier Number:  NBP1-74037G
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Antibody Type:
    Primary
  • Antigen Name:
    beta II Tubulin B
  • Clonality:
    Monoclonal
  • Clone:
    5B2
  • Conjugation:
    DyLight® 488
  • ELISA:
    Yes
  • Host:
    Mouse
  • ImmunoFluorescence:
    Yes
  • Isotype:
    IgG2b kappa
  • Reactivity:
    Human,
    Mouse
  • Western Blot:
    Yes
  • Size:
    100 μl
  • Gene ID:
    347733
  • Antigen Synonyms:
    DKFZp566F223|bA506K6.1|tubulin|tubulin beta-2B chain|MGC8685|beta 2B|FLJ98847|beta polypeptide paralog|class II beta-tubulin isotype
  • Storage Buffer:
    50mM Sodium Borate
  • Storage Temperature:
    Store at 4C in the dark.
  • Immunogen:
    Recombinant human beta-Tubulin (1-445aa) purified from E. coli
  • Purification:
    Protein G purified
  • Cat. No.:
    103402-766

Specifications

About this item

The beta II Tubulin B Antibody (5B2) [DyLight 488] from Novus Biologicals is a mouse monoclonal antibody to beta II Tubulin B. This antibody reacts with human, mouse. The beta II Tubulin B Antibody (5B2) [DyLight 488] has been validated for the following applications: Western Blot, ELISA, Immunocytochemistry / Immunofluorescence.

Type: Primary
Antigen: beta II Tubulin B
Clonality: Monoclonal
Clone: 5B2
Conjugation: DyLight 488
Epitope:
Host: Mouse
Isotype: IgG2b Kappa
Reactivity: Human, Mouse