To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Antibody Type:Primary
- Antigen Name:beta II Tubulin B
- Clonality:Monoclonal
- Clone:5B2
- Conjugation:Biotin
- ELISA:Yes
- Host:Mouse
- ImmunoFluorescence:Yes
- Isotype:IgG2b kappa
- Reactivity:Human,Mouse
- Western Blot:Yes
- Size:100 μl
- Environmentally Preferable:
- Gene ID:347733
- Antigen Synonyms:DKFZp566F223|bA506K6.1|tubulin|tubulin beta-2B chain|MGC8685|beta 2B|FLJ98847|beta polypeptide paralog|class II beta-tubulin isotype
- Storage Buffer:PBS
- Storage Temperature:Store at 4C in the dark.
- Immunogen:Recombinant human beta-Tubulin (1-445aa) purified from E. coli
- Purification:Protein G purified
- Cat. No.:103404-958
Specifications
About this item
The beta II Tubulin B Antibody (5B2) [Biotin] from Novus Biologicals is a mouse monoclonal antibody to beta II Tubulin B. This antibody reacts with human, mouse. The beta II Tubulin B Antibody (5B2) [Biotin] has been validated for the following applications: Western Blot, ELISA, Immunocytochemistry / Immunofluorescence.
Type: Primary
Antigen: beta II Tubulin B
Clonality: Monoclonal
Clone: 5B2
Conjugation: Biotin
Epitope:
Host: Mouse
Isotype: IgG2b Kappa
Reactivity: Human, Mouse