Anti-beta 2-Microglobulin Mouse Monoclonal Antibody (DyLight 650) [clone: SPM617]
103402-682
- Antibody Type:Primary
- Antigen Name:beta 2-Microglobulin
- Clonality:Monoclonal
- Clone:SPM617
- Conjugation:DyLight® 650
- Flow Cytometry:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgG1 kappa
- Reactivity:Primates,Human
- Size:100 μl
- Gene ID:567
- Antigen Synonyms:beta-2-microglobin|beta chain of MHC class I molecules|beta-2-microglobulin
- Storage Buffer:50mM Sodium Borate
- Storage Temperature:Store at 4C in the dark.
- Immunogen:Full length recombinant human B2M protein
- Purification:Protein A or G purified
- Cat. No.:103402-682
The beta 2-Microglobulin Antibody (SPM617) [DyLight 650] from Novus Biologicals is a mouse monoclonal antibody to beta 2-Microglobulin. This antibody reacts with human, primate. The beta 2-Microglobulin Antibody (SPM617) [DyLight 650] has been validated for the following applications: Flow Cytometry, Immunohistochemistry-Paraffin.
Type: Primary
Antigen: beta 2-Microglobulin
Clonality: Monoclonal
Clone: SPM617
Conjugation: DyLight 650
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human, Primate