Order Entry
United States
ContactUsLinkComponent
 
Anti-alpha-Smooth Muscle Actin Mouse Monoclonal Antibody (DyLight 405) [clone: 1A4 + ACTA2/791]
Catalog # 103410-668
Supplier:  Avantor
Anti-alpha-Smooth Muscle Actin Mouse Monoclonal Antibody (DyLight 405) [clone: 1A4 + ACTA2/791]
Catalog # 103410-668
Supplier:  Avantor
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Antibody Type:
    Primary
  • Antigen Name:
    alpha-Smooth Muscle Actin
  • Clonality:
    Monoclonal
  • Clone:
    1A4 + ACTA2/791
  • Conjugation:
    DyLight® 405
  • Flow Cytometry:
    Yes
  • Host:
    Mouse
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human,
    Rat
  • Size:
    100 μl
  • Environmentally Preferable:
  • Gene ID:
    59
  • Antigen Synonyms:
    aorta|alpha smooth muscle actin|smooth muscle|Cell growth-inhibiting gene 46 protein|AAT6|alpha 2|alpha-SMA|smooth muscle actin|sma|a-sma|ACTVS|alpha 2 actin|aortic smooth muscle|alpha-actin-2|actin|growth-inhibiting gene 46|alpha SMA
  • Storage Buffer:
    50mM Sodium Borate
  • Storage Temperature:
    Store at 4C in the dark.
  • Immunogen:
    N-Terminal decapeptide of alpha smooth muscle isoform of actin and conjugated to KLH (1A4); Recombinant full-length human ACTA2 protein (ACTA2/791)
  • Purification:
    Protein A or G purified
  • Cat. No.:
    103410-668

Specifications

About this item

The alpha-Smooth Muscle Actin Antibody (1A4 + ACTA2 / 791) [DyLight 405] from Novus Biologicals is a mouse monoclonal antibody to alpha-Smooth Muscle Actin. This antibody reacts with human, rat. The alpha-Smooth Muscle Actin Antibody (1A4 + ACTA2 / 791) [DyLight 405] has been validated for the following applications: Flow Cytometry, Immunohistochemistry-Paraffin.

Type: Primary
Antigen: alpha-Smooth Muscle Actin
Clonality: Monoclonal
Clone: 1A4 + ACTA2/791
Conjugation: DyLight 405
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human, Rat