Order Entry
United States
ContactUsLinkComponent
Anti-alpha-Smooth Muscle Actin Mouse Monoclonal Antibody (PerCP (Peridinin-Chlorophyll Protein Complex)) [clone: 1A4/asm-1]
Anti-alpha-Smooth Muscle Actin Mouse Monoclonal Antibody (PerCP (Peridinin-Chlorophyll Protein Complex)) [clone: 1A4/asm-1]
Catalog # 103412-988
Supplier:  Novus Biologicals
Anti-alpha-Smooth Muscle Actin Mouse Monoclonal Antibody (PerCP (Peridinin-Chlorophyll Protein Complex)) [clone: 1A4/asm-1]
Catalog # 103412-988
Supplier:  Novus Biologicals
Supplier Number:  NBP2-34522PCP

Specifications

  • Antibody Type:
    Primary
  • Antigen Name:
    alpha-Smooth Muscle Actin
  • Clonality:
    Monoclonal
  • Clone:
    1A4/asm-1
  • Conjugation:
    PerCP (Peridinin-Chlorophyll Protein Complex)
  • Flow Cytometry:
    Yes
  • Host:
    Mouse
  • ImmunoChemistry:
    Yes
  • ImmunoFluorescence:
    Yes
  • ImmunoPrecipitation:
    Yes
  • Isotype:
    IgG2a kappa
  • Reactivity:
    Rat,
    Mouse,
    Cat,
    Baboon,
    Pig,
    Rabbit,
    Monkey,
    Sheep,
    Goat,
    Chicken,
    Human,
    Bovine,
    Guinea pig,
    Dog
  • Western Blot:
    Yes
  • Size:
    100 μl
  • Gene ID:
    59
  • Antigen Synonyms:
    aorta|alpha smooth muscle actin|smooth muscle|Cell growth-inhibiting gene 46 protein|AAT6|alpha 2|alpha-SMA|smooth muscle actin|sma|a-sma|ACTVS|alpha 2 actin|aortic smooth muscle|alpha-actin-2|actin|growth-inhibiting gene 46|alpha SMA
  • Storage Buffer:
    PBS
  • Storage Temperature:
    Store at 4C in the dark.
  • Immunogen:
    N-Terminal decapeptide of alpha smooth muscle isoform of actin and conjugated to KLH
  • Purification:
    Protein A purified
  • Cat. No.:
    103412-988

Specifications

About this item

The alpha-Smooth Muscle Actin Antibody (1A4 / asm-1) [PerCP] from Novus Biologicals is a mouse monoclonal antibody to alpha-Smooth Muscle Actin. This antibody reacts with human, mouse, rat, porcine, baboon, bovine, canine, chicken, feline, guinea pig, goat, monkey, rabbit, sheep. The alpha-Smooth Muscle Actin Antibody (1A4 / asm-1) [PerCP] has been validated for the following applications: Western Blot, Flow Cytometry, Immunocytochemistry / Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.

Type: Primary
Antigen: alpha-Smooth Muscle Actin
Clonality: Monoclonal
Clone: 1A4/asm-1
Conjugation: PerCP (Peridinin-Chlorophyll Protein Complex)
Epitope:
Host: Mouse
Isotype: IgG2a Kappa
Reactivity: Human, Mouse, Rat, Porcine, Baboon, Bovine, Canine, Chicken, Feline, Guinea Pig, Goat, Monkey, Rabbit, Sheep