Anti-Adenosine A2a R Mouse Monoclonal Antibody (DyLight 755) [clone: 7F6-G5-A2]
Catalog # 103409-238
Supplier: Novus Biologicals
Specifications
- Antibody Type:Primary
- Antigen Name:Adenosine A2a R
- Clonality:Monoclonal
- Clone:7F6-G5-A2
- Conjugation:DyLight® 755
- Flow Cytometry:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG2a
- Reactivity:Pig,Rabbit,Human,Guinea pig,Dog,Rat,Mouse
- Western Blot:Yes
- Size:100 μl
- Epitope:Epitope mapped to SQPLPGER in the third intracellular loop (PMID 9822147).
- Gene ID:135
- Antigen Synonyms:adenosine A2 receptor|adenosine A2a receptor|adenosine receptor subtype A2a|ADORA2|hA2aR|adenosine receptor A2a|ADORA 2|RDC8
- Storage Buffer:50mM Sodium Borate
- Storage Temperature:Store at 4C in the dark.
- Immunogen:Full length recombinant human Adenosine A2a R. [UniProt# P29274]
- Purification:Protein G purified
- Cat. No.:103409-238
Specifications
About this item
The Adenosine A2a R Antibody (7F6-G5-A2) [DyLight 755] from Novus Biologicals is a mouse monoclonal antibody to Adenosine A2a R. This antibody reacts with human, mouse, rat, porcine, canine, guinea pig, rabbit. The Adenosine A2a R Antibody (7F6-G5-A2) [DyLight 755] has been validated for the following applications: Western Blot, Flow Cytometry, Immunocytochemistry / Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.
Type: Primary
Antigen: Adenosine A2a R
Clonality: Monoclonal
Clone: 7F6-G5-A2
Conjugation: DyLight 755
Epitope: Epitope mapped to SQPLPGER in the third intracellular loop (PMID 9822147).
Host: Mouse
Isotype: IgG2a
Reactivity: Human, Mouse, Rat, Porcine, Canine, Guinea Pig, Rabbit