Anti-CNX Mouse Monoclonal Antibody (DyLight 405) [clone: IE2.1C12]
Catalog # 103395-436
Supplier: Novus Biologicals
Specifications
- Antibody Type:Primary
- Antigen Name:Calnexin
- Antigen Symbol:CNX
- Clonality:Monoclonal
- Clone:IE2.1C12
- Conjugation:DyLight® 405
- Flow Cytometry:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG2b kappa
- Reactivity:Human
- Western Blot:Yes
- Size:100 μl
- Gene ID:821
- Antigen Synonyms:calnexin|Major histocompatibility complex class I antigen-binding protein p88|CNX|P90|IP90FLJ26570
- Storage Buffer:50mM Sodium Borate
- Storage Temperature:Store at 4C in the dark.
- Immunogen:Partial recombinant human Calnexin protein (between amino acids 1-300) [UniProt P27824]
- Purification:Protein G purified
- Cat. No.:103395-436
Specifications
About this item
The Calnexin Antibody (IE2.1C12) [DyLight 405] from Novus Biologicals is a mouse monoclonal antibody to Calnexin. This antibody reacts with human. The Calnexin Antibody (IE2.1C12) [DyLight 405] has been validated for the following applications: Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry / Immunofluorescence, Immunohistochemistry-Paraffin, Flow (Intracellular).
Type: Primary
Antigen: CNX
Clonality: Monoclonal
Clone: IE2.1C12
Conjugation: DyLight 405
Epitope:
Host: Mouse
Isotype: IgG2b Kappa
Reactivity: Human