To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
- Antibody Type:Primary
- Antigen Symbol:CD277/BTN3A1
- Clonality:Monoclonal
- Clone:BT3.1
- Conjugation:DyLight® 680
- Flow Cytometry:Yes
- Host:Mouse
- Isotype:IgG1
- Reactivity:Human
- Size:100 μl
- Gene ID:11119
- Storage Buffer:50mM Sodium Borate
- Storage Temperature:Store at 4C in the dark.
- Immunogen:The immunogen for this antibody is CD277 (B7 family, Butyrophilin).
- Purification:Protein A or G purified
- Cat. No.:103401-438
The CD277 / BTN3A1 Antibody (BT3.1) [DyLight 680] from Novus Biologicals is a mouse monoclonal antibody to CD277 / BTN3A1. This antibody reacts with human. The CD277 / BTN3A1 Antibody (BT3.1) [DyLight 680] has been validated for the following applications: Flow Cytometry.
Type: Primary
Antigen: CD277/BTN3A1
Clonality: Monoclonal
Clone: BT3.1
Conjugation: DyLight 680
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human