Anti-CD2 Mouse Monoclonal Antibody (DyLight 488) [clone: 1E7E8.G4]
Catalog # 103397-926
Supplier: Novus Biologicals
Specifications
- Antibody Type:Primary
- Antigen Symbol:CD2
- Clonality:Monoclonal
- Clone:1E7E8.G4
- Conjugation:DyLight® 488
- ELISA:Yes
- Flow Cytometry:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG2a kappa
- Reactivity:Human
- Western Blot:Yes
- Size:100 μl
- Gene ID:914
- Antigen Synonyms:sheep red blood cell receptor|Rosette receptor|CD2 molecule|Erythrocyte receptor|T-cell surface antigen T11/Leu-5|T11|T-cell surface antigen CD2|SRBC|LFA-3 receptor|CD2 antigen (p50)|lymphocyte-function antigen-2|CD2 antigen|LFA-2|FLJ46032
- Storage Buffer:50mM Sodium Borate
- Storage Temperature:Store at 4C in the dark.
- Immunogen:Human Thymocytes/Sezary T cells
- Purification:Protein A purified
- Cat. No.:103397-926
Specifications
About this item
The CD2 Antibody (1E7E8.G4) [DyLight 488] from Novus Biologicals is a mouse monoclonal antibody to CD2. This antibody reacts with human. The CD2 Antibody (1E7E8.G4) [DyLight 488] has been validated for the following applications: Western Blot, Flow Cytometry, ELISA, Immunocytochemistry / Immunofluorescence, Immunohistochemistry-Frozen.
Type: Primary
Antigen: CD2
Clonality: Monoclonal
Clone: 1E7E8.G4
Conjugation: DyLight 488
Epitope:
Host: Mouse
Isotype: IgG2a Kappa
Reactivity: Human