Anti-CCL8/MCP-2 Mouse Monoclonal Antibody (FITC (Fluorescein Isothiocyanate)) [clone: 35509.11]
Catalog # 103397-306
Supplier: Novus Biologicals
Specifications
- Antibody Type:Primary
- Antigen Symbol:CCL8/MCP-2
- Clonality:Monoclonal
- Clone:35509.11
- Conjugation:FITC (Fluorescein Isothiocyanate)
- ELISA:Yes
- Host:Mouse
- Isotype:IgG1
- Reactivity:Human,Mouse
- Size:100 μl
- Gene ID:6355
- Antigen Synonyms:HC14|member 8 (monocyte chemotactic protein 2)|MCP2|MCP-2|monocyte chemoattractant protein 2|chemokine (C-C motif) ligand 8|C-C motif chemokine 8|SCYA10|small-inducible cytokine A8
- Storage Buffer:PBS
- Storage Temperature:Store at 4C in the dark.
- Immunogen:Recombinant human monocyte chemotactic protein 2 (MCP2), expressed in E. coli.
- Purification:Protein A purified
- Cat. No.:103397-306
Specifications
About this item
The CCL8 / MCP-2 Antibody (35509.11) [FITC] from Novus Biologicals is a mouse monoclonal antibody to CCL8 / MCP-2. This antibody reacts with human, mouse. The CCL8 / MCP-2 Antibody (35509.11) [FITC] has been validated for the following applications: ELISA.
Type: Primary
Antigen: CCL8/MCP-2
Clonality: Monoclonal
Clone: 35509.11
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse