Order Entry
United States
ContactUsLinkComponent
Anti-CASK Rabbit Polyclonal Antibody
Anti-CASK Rabbit Polyclonal Antibody
Catalog # 102706-516
Supplier:  Novus Biologicals
Anti-CASK Rabbit Polyclonal Antibody
Catalog # 102706-516
Supplier:  Novus Biologicals
Supplier Number:  NBP2-15710
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Antibody Type:
    Primary
  • Antigen Symbol:
    CASK
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • ImmunoFluorescence:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human,
    Mouse
  • Western Blot:
    Yes
  • Size:
    100 μl
  • Environmentally Preferable:
  • Gene ID:
    8573
  • Antigen Synonyms:
    Protein lin-2 homolog|CMG|calcium/calmodulin-dependent serine protein kinase membrane-associatedguanylate kinase|FGS4CAGH39|calcium/calmodulin-dependent serine protein kinase (MAGUK family)|TNRC8|hCASK|FLJ22219|EC 2.7.11.1|Calcium/calmodulin-dependent serine protein kinase|EC 2.7.11|trinucleotide repeat containing 8|MICPCH|peripheral plasma membrane protein CASK|CAMGUK|LIN2FLJ31914
  • Storage Buffer:
    0.1M Tris (pH 7.0), 0.1M Glycine and 10% Glycerol
  • Storage Temperature:
    Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
  • Concentration:
    1 mg/ml
  • Immunogen:
    Recombinant protein encompassing a sequence within the center region of human CASK. The exact sequence is proprietary.
  • Purification:
    Immunogen affinity purified
  • Cat. No.:
    102706-516
  • Supplier no.:
    NBP2-15710

Specifications

About this item

The CASK Antibody from Novus Biologicals is a rabbit polyclonal antibody to CASK. This antibody reacts with human, mouse. The CASK Antibody has been validated for the following applications: Western Blot, Immunocytochemistry / Immunofluorescence.

Type: Primary
Antigen: CASK
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse