Anti-CART1 Mouse Monoclonal Antibody [clone: 2A10]
Catalog # 103334-676
Supplier: Novus Biologicals
Specifications
- Antibody Type:Primary
- Antigen Symbol:CART1
- Clonality:Monoclonal
- Clone:2A10
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- ImmunoFluorescence:Yes
- Isotype:IgG2a kappa
- Reactivity:Human
- Western Blot:Yes
- Size:100 μg
- Gene ID:8092
- Antigen Synonyms:ALX homeobox 1|cartilage paired-class homeoprotein 1|CART-1|CART1ALX homeobox protein 1|FND3|Cartilage homeoprotein 1
- Storage Buffer:PBS (pH 7.4)
- Storage Temperature:Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
- Immunogen:CART1 (NP_008913 198 a.a. - 306 a.a.) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. QAKSHFAATYDISVLPRTDSYPQIQNNLWAGNASGGSVVTSCMLPRDTSSCMTPYSHSPRTDSSYTGFSNHQNQFSHVPLNNFFTDSLLTGATNGHAFETKPEFERRSS
- Purification:IgG purified
- Cat. No.:103334-676
- Supplier no.:H00008092-M02
Specifications
About this item
The CART1 Antibody (2A10) from Novus Biologicals is a mouse monoclonal antibody to CART1. This antibody reacts with human. The CART1 Antibody (2A10) has been validated for the following applications: Western Blot, ELISA, Immunocytochemistry / Immunofluorescence.
Type: Primary
Antigen: CART1
Clonality: Monoclonal
Clone: 2A10
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a Kappa
Reactivity: Human